To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:
- Size:25 μg
- Environmentally Preferable:
- Storage Conditions:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70 °C as supplied. 3 months, -70 °C under sterile conditions after opening.
- Endotoxin Content:<0.01 EU per 1 ╡g of the protein by the LAL method.
- Gene ID:55577
- Protein Synonyms:GlcNAc Kinase|N-acetylglucosamine kinaseHSA242910|N-Acetylglucosamine Kinase|EC 2.7.1.59|N-acetyl-D-glucosamine kinase|GNK|GNKGlcNAc kinase
- Accession No.:P75959
- Protein/Peptide Name:N-Acetyl-D-Glucosamine Kinase/NAGK
- Purity:>90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie« Blue stain at 5 ╡g per lane
- Molecular Weight:37-38 kDa, reducing conditions
- Cat. No.:102869-222
- Supplier no.:8020-GK-025
The Recombinant E. coli N-Acetyl-D-Glucosamine Kinase/NAGK from R&D Systems is derived from E. coli. The Recombinant E. coli N-Acetyl-D-Glucosamine Kinase/NAGK has been validated for the following applications: Enzyme Activity.