Human Recombinant CD42b/GPIb alpha (from NS0 Cells)
Catalog # 220024-520
Supplier: R&D Systems
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:NS0 cells
- Species:Human
- Size:50 μg
- Environmentally Preferable:
- Storage Conditions:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
- Endotoxin Content:<0.10 EU per 1 ╡g of the protein by the LAL method.
- Gene ID:2811
- Protein Synonyms:CD42b|CD42b antigen|GP1B|Antigen CD42b-alpha|CD42b-alpha|GPIb alpha|platelet glycoprotein Ib alpha chain|glycoprotein Ib (platelet)|platelet membrane glycoprotein 1b-alpha subunit|GP-Ib alpha|BSS|GPIbA|GPIb-alpha|GP1BA|MGC34595|BP1BA|alpha polypeptide|Glycoprotein Ibalpha
- Accession No.:P07359.1
- Protein/Peptide Name:CD42b/GPIb alpha
- Purity:>90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
- Molecular Weight:95-120 kDa, reducing conditions
- Cat. No.:220024-520
- Supplier no.:4067-GP-050
Specifications
About this item
The Recombinant Human CD42b/GPIb alpha Protein from R&D Systems is derived from NS0. The Recombinant Human CD42b/GPIb alpha Protein has been validated for the following applications: Binding Activity.