Human Recombinant DPPII/QPP/DPP7 (from NS0 Cells)
Catalog # 220024-245
Supplier: R&D Systems
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:NS0 cells
- Species:Human
- Size:10 μg
- Environmentally Preferable:
- Storage Conditions:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70 °C as supplied. 3 months, -20 to -70 °C under sterile conditions after opening.
- Endotoxin Content:<1.0 EU per 1 ╡g of the protein by the LAL method.
- Gene ID:29952
- Protein Synonyms:QPP|dipeptidyl-peptidase 7|dipeptidyl arylamidase II|DPP2|DPP II|DPP7|carboxytripeptidase|Dipeptidyl aminopeptidase II|EC 3.4.14.2|DPPII|Dipeptidyl peptidase II|dipeptidylpeptidase 7|dipeptidyl peptidase 2|Quiescent cell proline dipeptidase|dipeptidyl-peptidase II|Dipeptidyl peptidase 7
- Accession No.:AAH11907
- Protein/Peptide Name:DPPII/QPP/DPP7
- Purity:>95%, by SDS-PAGE under reducing conditions and visualized by silver stain
- Molecular Weight:64 kDa, reducing conditions
- Cat. No.:220024-245
- Supplier no.:3438-SE-010
Specifications
About this item
The Recombinant Human DPPII/QPP/DPP7 Protein from R&D Systems is derived from NS0. The Recombinant Human DPPII/QPP/DPP7 Protein has been validated for the following applications: Enzyme Activity.