To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Description:Amylo-Glo RTD amyloid plaque stain reagent 100X
- Size:5 ml
- Environmentally Preferable:
- ImmunoFluorescence:Yes
- Storage Temperature:The stock solution can be stored for up to 6 months after date of receipt at 4 °C protected from light. No preservatives. Use sterile technique when handling and proper laboratory procedures.
- Concentration:100X
- Formulation:Liquid
- Shipping Temperature:Ambient
- Cat. No.:76264-656
Amylo-Glo® RTD™ 'Ready to Dilute' Staining reagent is designed to stain amyloid plaques in tissue sections.
- For Amyloid plaques - Both intraneuronal and vascular
- Very bright blue UV excitable stain under physiological conditions
- Unique excitation/emission profile and mild staining
- Combination for multiple immunofluorescent labeling studies
- Storage Up to 6 months after date of receipt at 4 °C protected from light
This novel marker has several advantages over other conventional markers such as Thioflavin S and Congo Red because of its unique chemical and spectral properties. (L. Schmued et al. (2012) J.Neuroscience Methods 209:120– 126). Using Amylo-Glo® results in a very bright blue UV excitable stain under physiological conditions that will not bleed through when illuminated with other filters. Its brightness makes it ideal for low magnification quantification studies, while its unique excitation/emission profile and mild staining conditions makes it ideal for combination for multiple immunofluorescent labeling studies.
Amylo-Glo® RTD™ is compatible with fresh, frozen, and formalin-fixed immunohistochemistry or cytochemistry, and it is particularly good for confocal and multiple labeling because of its high fluorescent intensity and high resistance to photo-bleaching. Moreover because Amylo-Glo® fluoresces in the UV channel, double and triple labeling experiments can be performed very easily (see protocol). Reagent detects amyloid plaques, both intraneuronal and vascular.