About this item
Produced from sera of rabbits immunized with highly pure Recombinant Murine MIP-2 (CXCL2). AntiMurine MIP-2 (CXCL2)specific antibody was purified by affinity chromatography and then biotinylated.
Applications tested can vary by lot number. See Product Data Sheet for more information.
Type: Secondary
Antigen: MIP-2 (CXCL2)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Murine
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 10776-268
- 10776-270
- Antigen Symbol:
- MIP-2 (CXCL2)
- MIP-2 (CXCL2)
- Antigen Name:
- MIP-2
- MIP-2
- Antibody Type:
- Secondary
- Secondary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Mouse
- Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Murine MIP-2 (CXCL2)
- Recombinant Murine MIP-2 (CXCL2)
- Form:
- Lyophilized
- Lyophilized
- Molecular Weight:
- 7.8 kDa
- 7.8 kDa
- Purification:
- Recombinant Murine MIP-2 (CXCL2)
- Recombinant Murine MIP-2 (CXCL2)
- Storage Buffer:
- See Product Data Sheet for more information.
- See Product Data Sheet for more information.
- Storage Temperature:
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Shipping Temperature:
- Ambient
- Ambient