Specifications
- Size:1 L
- Environmentally Preferable:
- Storage Temperature:See product insert for storage instructions.
- Cat. No.:10771-876
- Supplier no.:AFCHO
Specifications
About this item
PeproTech's PeproGrow™ AF-CHO is an animal component-free, serum-free, complete medium formulation for the in vitro cultivation of Chinese Hamster Ovary-S cells.
- Complete Media
- Animal component-Free
- Serum-Free
- Protein-Free
- High recombinant protein expression
PeproGrow™ CD-CHO is an animal component-free, serum-free, protein-free, chemically-defined, complete medium formulation for the in vitro cultivation of Chinese Hamster Ovary-S cells (Thermo Fisher Scientific catalog numbers 11619-012, R800-07, or A11557-01). This medium is intended for recombinant protein expression in suspension culture. This complete medium contains L-alanyl-L-Glutamine, amino acids, vitamins, and salts, which is recommended for a 5 day batch culture with a seeding density of 0.5 x 106 cells/mL. An adaptation process is not required for Thermo Fisher Scientific CHO-S cells.