ContactUsLinkComponent
Left Handed Mayo Scissors, OR-Grade, Sklar®
Left Handed Mayo Scissors, OR-Grade, Sklar®
Catalog # 76191-388
Supplier:  Sklar
undefined
Left Handed Mayo Scissors, OR-Grade, Sklar®
Catalog # 76191-388
Supplier:  Sklar
Supplier Number:  15-1555LFT
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Left Handed Mayo Scissors
  • Length:
    5.5"
  • Material:
    Stainless steel
  • Environmentally Preferable:
  • Curvature:
    Straight
  • Cat. No.:
    76191-388
Specifications
About this item
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Mayo Dissecting Scissors are a versatile instrument that can be used in multiple procedures and settings. They have a standard beveled blade. While the curved instrument is most commonly utilized for cutting or dissecting dense or deep tissue, and the straight version is typically used for cutting surface tissue or sutures, both may be used interchangeably depending on procedure and physician preference. Sklar® also offers Mayo Dissecting Scissors with a selection of special features to meet procedure specifications and surgeon preferences.