About this item
All Sklar products are industry standard.
- For more than 120 years, Sklar has set the standard for surgical instrumentation
- With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Miller Laryngoscope Blades are used for direct, rigid laryngoscopy procedures to view vocal cords, remove foreign objects from the throat, biopsy, or tracheal intubation. The Miller blade is straight and therefore longer than the curved Macintosh blade. It is inserted past the epiglottis, typically causing less trauma during intubation. The Miller blade is commonly used with pediatric and infant patients.