About this item
The CryoStore™ bag is designed to cryogenically freeze blood components. This product is for single use only.
- Does not contain phthalates, BPA, or latex
- Needle-free injection port
- Heat welded permanent lot number into the tail of each bag for easy traceability
- Hermetically sealed spike ports and film are made of rugged EVA for better durability
Each bag has a donor tube with printed segments of approximately 100µL and a matched number on the bag for keeping aliquot samples with the bag. The donor tube is intentionally placed in the corner of the bag for easier de-bubbling and sealing.