ContactUsLinkComponent
ATCC® CRL-3216™ 293T Cell Line, American Type Culture Collection
ATCC® CRL-3216™ 293T Cell Line, American Type Culture Collection
Catalog # MSPP-CRL3216
Supplier:  0000013421
CAS Number:  
ATCC® CRL-3216™ 293T Cell Line, American Type Culture Collection
Catalog # MSPP-CRL3216
Supplier:  0000013421
Supplier Number:  CRL-3216
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    293T Cell Line
  • Environmentally Preferable:
  • Cat. No.:
    MSPP-CRL3216
  • Supplier no.:
    CRL-3216
Specifications
About this item
The 293T cell line, originally referred as 293tsA1609neo, is a highly transfectable derivative of human embryonic kidney 293 cells, and contains the SV40 T-antigen.
This cell line is competent to replicate vectors carrying the SV40 region of replication. It gives high titers when used to produce retroviruses. It is widely used for retroviral production, gene expression and protein production.
Caution: Intended for research use only. It is not intended for any animal or human therapeutic or diagnostic use.