Order Entry
United States
ContactUsLinkComponent
EnzyFluo™ Myeloperoxidase Assay Kit, BioAssay Systems
EnzyFluo™ Myeloperoxidase Assay Kit, BioAssay Systems
Catalog # 76236-154
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
undefined
EnzyFluo™ Myeloperoxidase Assay Kit, BioAssay Systems
Catalog # 76236-154
Supplier:  BIOASSAY SYSTEMS
Supplier Number:  EMPO-100
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    EnzyFluo™ Myeloperoxidase Assay Kit
  • Size:
    100 assays
  • Environmentally Preferable:
  • Cat. No.:
    76236-154

Specifications

About this item

MPO peroxidation activity determination in biological samples (example: cell lysates, tissues, and more.)

  • Fast and sensitive
  • Linear detection range (20 µl sample): 0.0025 to 2 U/L for 10 min reaction at 25 °C.
  • Convenient and high-throughput, homogeneous "mix-incubate-measure" type assay
  • Can be readily automated on HTS liquid handling systems for processing thousands of samples per day
  • Method: FL530/585 nm
  • Samples: Cell lysates, tissues,and more

Myeloperoxidase (MPO; EC 1.11.2.2) is a peroxidase enzyme and can be found in neutrophil, monocytes, and some soft tissue macrophages. MPO has an ability to use chloride as a cosubstrate with hydrogen peroxide to generate hypochlorous acid, a powerful antimicrobial agent produced by neutrophils. However, an excessive production of hypochlorous acid can lead to oxidative stress and tissues damage. Inflammation may also result when MPO oxidizes various substances such as phenols and anilines. Studies show that increased MPO levels may increase the risk of myocardial infarction and cardiovascular disease.

EnzyFluoTM myeloperoxidase (MPO) assay kit is based on the MPO enzyme reaction with hydrogen peroxide (H₂O₂) which oxidizes the dye reagent to a highly fluorescent product. The fluorescence intensity of this product, measured at &labda;ex/em = 530/585 nm, is proportional to the total peroxidation activity in the sample. The provided MPO inhibitor is used to suppress peroxidase activity due to MPO in order to differentiate other peroxidase activities that may be present in the samples.