Order Entry
United States
ContactUsLinkComponent
ATCC® RPMI-1640 Medium, American Type Culture Collection
ATCC® RPMI-1640 Medium, American Type Culture Collection
Catalog # MSPP-302001
Supplier:  0000013421
CAS Number:  
ATCC® RPMI-1640 Medium, American Type Culture Collection
Catalog # MSPP-302001
Supplier:  0000013421
Supplier Number:  30-2001
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    RPMI-1640 Medium, pH 7.0 – 7.4
  • Size:
    500 ml
  • Environmentally Preferable:
  • Cat. No.:
    MSPP-302001
  • Supplier no.:
    30-2001

Specifications

About this item

RPMI1640 medium modified to contain 2 mM Lglutamine, 10 mM HEPES, 1 mM sodium pyruvate, 4500 mg/L glucose, and 1500 mg/L sodium bicarbonate, for use in incubators using 5% CO2 in air. Additional sodium bicarbonate may be required for use in incubators containing higher percentages of CO2.

  • Osmolality: 246 to 306 mOsm/kg H2O
  • Endotoxin: ≤0.5 EU/mL
  • Storage: 2 °C to 8 °C

Sterility tests are performed on each lot of medium using current USP methods. Each lot of medium is also checked for its ability to support the growth of several different cell lines using both sequential subcultures and plating efficiencies.