Laboratory Glassware Drying Ovens, Yamato
Supplier: Yamato Scientific
Certificates
About this item
Glassware Drying Ovens featuring a large window for easy observation.
- Can be used to store instruments after drying
- Highly efficient heat insulation material for both internal and external structure
- Adjustable foot for stability on uneven floors
- Mobile on casters (Models DG800C/810C/840C/850C)
- Equipped with stainless steel pipe heater and water receiving plate at the bottom
- Stainless steel interior, easy to clean and highly resistant to corrosion
- Models DG440C/450C/840C/850C installed with filter at air in-take port, exhaust fan and germicidal lamp for fast drying
- Dial setting and digital display of temperature control and timer
The ovens have a standard-glass window, a door, and a stainless steel interior that is easy to clean, resistant to corrosion, and works as instrument storage after drying.
The heat insulation material makes the internal and external structure even more efficient. To ensure safe operation, self-diagnostic functions, independent overheat prevention, and an over-current leakage breaker are provided. Additional functions include calibration offset, power failure operation, key lock, and auto start and stop.
Specifications
- Catalog No:
- 76209-654
- 76209-652
- 76209-726
- 76209-658
- 76209-724
- 76209-656
- 76209-698
- 76209-696
- Control Type:
- Digital PID Controller
- Digital PID Controller
- Digital PID Controller
- Digital PID Controller
- Digital PID Controller
- Digital PID Controller
- Digital PID Controller
- Digital PID Controller
- Exhaust ports:
- 2 pieces
- 2 pieces
- 2 pieces
- —
- 2 pieces
- —
- —
- —
- No. of shelf rest steps/pitch:
- 10 Steps/30 mm
- 10 Steps/30 mm
- 29 Steps/30 mm
- 10 Steps/30 mm
- 29 Steps/30 mm
- 10 Steps/30 mm
- 29 Steps/30 mm
- 29 Steps/30 mm