About this item
Vector Laboratories horseradish peroxidase (HRP) substrate kits include options that are cost-effective (e.g. standard DAB), sensitive (e.g ImmPACT DAB) and convenient (ImmPACT DAB EqV). The range of vibrant colors facilitates multiplex staining and improved contrast with pigmented tissues and counterstains.
- Optimized for IHC, ICC, ISH and blots
- Stock solutions are supplied in convenient dropper bottles for greater ease of handling
- Compatible with either VectaMount Permanent Mounting Media (H-6000) or VectaMount AQ Aqueous Mounting Media (H-5501)
- One year expiry date
For immuno-visualization experiments to be successful, one needs proper sample prep, a well-targeted primary anybody, an effective secondary antibody-based detection system and a high-quality chromogenic substrate. Vector Laboratories has decades of experience offering exceptional staining systems and substrates.
Standard substrates provide exceptional value and convenience over substrates in the form of powders or pills and work well for higher-abundance targets. Vector Laboratories ImmPACT substrates offer up to five times higher sensitivity with convenient handling and are ideal for detecting lower-abundance targets.