Specifications
- Accuracy:2 - 3%
- Description:Automated flow injection system
- Environmentally Preferable:
- Width:16.5 cm
- Depth:24 cm
- Height:24 cm
- Weight:6.75 kg
- Cat. No.:101169-268
- Supplier no.:FIA-LAB-2500
Specifications
About this item
The flow injection analysis system was designed for automation of common environment and agricultural assay such as nitrate, phosphate and ammonia.
- Affordability and flexibility make the system ideal for teaching, research and commercial labs
- Easy to use
- Wavelength selection are made through software, no additional filters or lamps are required
It includes automated flow injection system with a four-channel peristaltic pump, a six-port injector valve with fittings and tubing, a flow cell, an LED light source and software.
Nitrate/Nitrite assay: This method performs FIA assays for soil testing and analysis of potable water and ground, surface, domestic and industrial wastewaters.
Phosphate assay: A phosphate measurement method based on U.S EPA protocols, best suited for agricultural and environmental testing.
Ammonia assay: An FIA assay for low ammonia concentrations using the salicylate method. Ideal for agricultural and environmental testing.