ContactUsLinkComponent
Aromatase (CYP19A) Activity Assay Kit (Fluorometric), BioVision
Aromatase (CYP19A) Activity Assay Kit (Fluorometric), BioVision
Catalog # 76211-954
Supplier:  Avantor
CAS Number:  
undefined
Aromatase (CYP19A) Activity Assay Kit (Fluorometric), BioVision
Catalog # 76211-954
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Aromatase (CYP19A) Activity Assay Kit (Fluorometric)
  • Size:
    100 assays
  • Environmentally Preferable:
  • Cat. No.:
    76211-954
Specifications
About this item
Highly Sensitive; Includes human CYP2C19 - NADPH Reductase as positive control.
  • Simple, convenient, highly sensitive
  • Fluorescent assay enables easy determination of Aromatase activity in a variety of biological samples
  • The substrate shows low background and a high signal-to-noise ratio
  • Kit includes Aromatase inhibitor, Letrozole, and a stable, recombinant human Aromatase as positive control
Aromatase (CYP19A, EC 1.14.14.14) is a member of the cytochrome P450 monooxidase (CYP) family of microsomal xenobiotic metabolism enzymes. Aromatase plays a critical role in steroidogenesis, catalyzing the conversion of androgenic hormones into estrogens. The enzyme is expressed in high levels in reproductive tissues, placenta, brain and adipose tissue and is responsible for mammalian sexual dimorphism and development of secondary sexual characteristics. Inhibitors of aromatase are used to treat estrogen-dependent breast cancer, as estrogens promote the expression of peptide growth factors responsible for tumorigenesis. Aromatase activity and expression can be affected by many organic environmental pollutants such as pesticides and plasticizers. Such compounds, known as endocrine disruptors, are suspected of causing precocious puberty, obesity, infertility and various cancers. BioVision’s Aromatase Activity Assay Kit enables rapid measurement of native or recombinant aromatase activity in biological samples such as placental microsomes. The assay utilizes a fluorogenic substrate that is converted into a highly fluorescent metabolite detected in the visible range (Ex/Em = 488/527 nm), ensuring a high signal-to-background ratio with little interference by autofluorescence. A highly selective aromatase inhibitor is provided for determination of aromatase activity in heterogeneous biological samples, where other CYP isozymes may contribute to substrate metabolism. The inhibitor displays greater than 100-fold selectivity for aromatase over other enzymes, ensuring targeted inhibition. Aromatase specific activity is calculated by running parallel reactions in the presence and absence of the inhibitor and subtracting any residual activity detected with the inhibitor present. The kit contains a complete set of reagents sufficient for 50 sets of paired reactions (in the presence and absence of inhibitor) and can detect a minimum of 25 nU aromatase activity.