ContactUsLinkComponent
EZScreen™ Glycogen Colorimetric Assay Kit, BioVision
EZScreen™ Glycogen Colorimetric Assay Kit, BioVision
Catalog # 76211-918
Supplier:  Avantor
CAS Number:  
undefined
EZScreen™ Glycogen Colorimetric Assay Kit, BioVision
Catalog # 76211-918
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    EZScreen™ Glycogen Colorimetric Assay Kit
  • Size:
    400 assays
  • Environmentally Preferable:
  • Cat. No.:
    76211-918
  • No. of wells:
    383 wells
Specifications
About this item
Highly Sensitive, minimum sample preparation required
  • Simple procedure; takes ~ 2 hours
  • Fast and convenient
  • Kit contains all necessary reagents for accurate measurement of Glycogen
Glycogen is a branched polysaccharide, made of glucose units that serves as the primary source of energy storage in animals. Most of the glucose residues in glycogen are linked by α-1,4-glycosidic bonds with branching via α-1,6 linkage. In mammals, the two major sites of glycogen storage are liver and skeletal muscle. In liver, glycogen synthesis and degradation are regulated to maintain blood-glucose levels, while stored glycogen in muscle can be processed to meet the energy needs of the muscle itself. Abnormal ability to utilize glycogen is found in patients suffering diabetes and other genetic glycogen storage related diseases. BioVision EZScreenTM Glycogen Colorimetric Assay Kit is an easy and convenient assay to measure glycogen levels in biological samples. In the assay, glucoamylase hydrolyzes the glycogen releasing glucose which is then specifically oxidized to produce a product that reacts with OxiRed Probe to generate color (OD 590 nm). The method is quantitative, rapid, simple, sensitive, and designed for high throughput format. The assay can detect as low as 0.025 µg of glycogen in 384-well assay format.