Order Entry
United States
ContactUsLinkComponent
Bacterial Genomic DNA Isolation Kit, BioVision, Inc.
Bacterial Genomic DNA Isolation Kit, BioVision, Inc.
Catalog # 76211-436
Supplier:  Avantor
CAS Number:  
undefined
Bacterial Genomic DNA Isolation Kit, BioVision, Inc.
Catalog # 76211-436
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Application:
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Bacterial DNA
  • Sample type:
    Bacteria
  • Target molecule:
    DNA
  • Description:
    Bacterial Genomic DNA Isolation Kit
  • Size:
    100 isolations
  • Environmentally Preferable:
  • Description:
    Bacterial Genomic DNA Isolation Kit
  • Cat. No.:
    76211-436

Specifications

About this item

Convenient and simple step-by-step method for isolating quality genomic DNA

  • Simple & rapid method to isolate highly pure, intact DNA from bacteria

Bacteria are one of the most abundant and diverse organisms on the planet, which take part in numerous critical ecosystem processes. Many bacterial species are pathogens that are responsible for causing a variety of human and animal diseases. In addition to their medical and ecological importance, bacteria are also used in various industrial applications such as production of enzymes and biofuels. BioVision’s bacterial genomic DNA isolation kit provides convenient and simple step-by-step method for isolating quality genomic DNA from gram-negative and gram-positive bacterial species. This kit utilizes enzymatic reactions to release bacterial DNA from the cell. DNA release from the cell is coupled with adsorption of DNA onto a silica spin-column in the presence of high salt concentration, eliminating the use of toxic organic compounds or solvents. DNA purified by this kit is suitable for various downstream molecular biology applications such as PCR, cloning, DNA hybridization, and Southern Blotting