Dispensing Bottle with Sealer Cap, LDPE, Dynalon
Catalog # 76210-726
Supplier: Dynalon
Specifications
- Cap Size:28 mm
- Ø:79 mm
- Capacity:500 ml (16 oz.)
- Height:195 mm
- Environmentally Preferable:
- Cat. No.:76210-726
- Weight:0.128 lbs.
Specifications
About this item
These low density polyethylene bottles are supplied with a sealer cap that snaps on easily after use and seals tightly.
- Maximum temperature for LDPE is 80 °C
- Offers good chemical resistance
- Suitable for mild and strong buffers
Spout openings are standard 0.030", and can be enlarged by cutting back with scissors.