ContactUsLinkComponent
Vitamin B12 Immunoaffinity Column, Eagle Biosciences, Inc.
Vitamin B12 Immunoaffinity Column, Eagle Biosciences, Inc.
Catalog # 102994-016
Supplier:  Eagle Biosciences
Vitamin B12 Immunoaffinity Column, Eagle Biosciences, Inc.
Catalog # 102994-016
Supplier:  Eagle Biosciences
Supplier Number:  BTCA318005
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Vitamin B12 Immunoaffinity Column, 10 columns
  • Environmentally Preferable:
  • Cat. No.:
    102994-016
  • Supplier no.:
    BTCA318005
Specifications
About this item
The Eagle Bio Vitamin B12 Immunoaffinity Column detects Vitamin B12 by combination of IAC and HPLC.
  • Sensitivity: 0.01 μg
  • Dynamic range: 0.01 to 5 μg
  • Sample size: 10 columns
Many methods of vitamin B12 determination are based on HPLC-UV (high performance liquid chromatography-ultraviolet) detection show low selectivity if problematic matrices are applied. This method of content determination of Vitamin B12 combines the high selectivity of immunoaffinity columns with its potential to concentrate, elute, and purify with the HPLC column.