Specifications
- Length:8.25"
- Material:Stainless Steel with Tungsten Carbide
- Environmentally Preferable:
- Curvature:Curved
- Cat. No.:76192-686
Specifications
About this item
The Sklar® Tungsten Carbide Julian needle holder is a ratcheted finger ring instrument used for suturing.
- For more than 120 years, Sklar has set the standard for surgical instrumentation
- With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
- All Sklar products are industry standard
The the pyramid shaped teeth ensure a firm grip withough slippage. TC instruments are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar® permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. These instruments are identified by gold handles.