ContactUsLinkComponent
PREVI® Color Gram, BioMerieux
PREVI® Color Gram, BioMerieux
Catalog # 76204-648
Supplier:  bioMerieux
CAS Number:  
PREVI® Color Gram, BioMerieux
Catalog # 76204-648
Supplier:  bioMerieux
Supplier Number:  414292
CAS Number:  
Specifications
  • Description:
    PREVI® Color Gram
  • Environmentally Preferable:
  • Cat. No.:
    76204-648
Specifications
About this item
Fully automated stain process from fixation to drying.
Includes PREVI® Color Gram Accessory Kit and PREVI® Color Gram Main Kit:
PREVI® Color Gram Accessory Kit (Contains 12 Slide Carosel, Waste Tubing, Waste Container V2, and Maintenance Kit V2)
PREVI® Color Gram Main Kit (Contains Priming Tool, Silicon Grease, Nozzle Wire, Nozzle Cleaning Strainer, Nozzles, Nozzle Tool, Nozzle Wrench, Nozzle Brush and Nozzle Cleaning Tubes)
Ordering information: It includes user manual, nozzle cleaning solution, D & E empty bottle, Color 12 acessory kit, Barcode.