To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Host:
- Primary antibody reactivity:
- Environmentally Preferable:
- Cat. No.:102650-528
- Supplier no.:F410
Specifications
About this item
This kit was developed using broadly reactive polyclonal antibodies to the hundreds of different HCPs and is intended for use in determining the presence of E.coli host cell protein contamination in products manufactured by recombinant expression.
Assay validation has determined that the LOD for this kit is ~0.2 ng/mL.