To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Source:
- Species:
- Size:10000 U
- Environmentally Preferable:
- Storage Conditions:–30...–10 °C
- Enzyme Name:S1 Nuclease
- Concentration:20 - 100 units/µl
- Tested Applications:Mapping, Duplex Shortening
- Cat. No.:PAM5761
- Supplier no.:M5761
Specifications
About this item
Endonuclease specific for single-stranded RNA or DNA.
- For mapping and duplex shortening
- Endonuclease Specific for Single-Stranded DNA and RNA
- Enzyme yields 5´-phosphoryl-terminated products
- Double-stranded nucleic acids (DNA:DNA, DNA:RNA or RNA:RNA) resist degradation except with extremely high concentrations of enzyme
- Supplied at a concentration of 20-100u/microl
S1 Nuclease degrades single-stranded DNA and RNA endonucleolytically to yield 5'-phosphoryl-terminated products. Double-stranded nucleic acids (DNA:DNA, DNA:RNA or RNA:RNA) are resistant to degradation except with extremely high concentrations of enzyme. The enzyme is used to remove single-stranded termini from double-stranded DNA, for selective cleavage of single-stranded DNA and for mapping RNA transcripts.