Order Entry
United States
ContactUsLinkComponent
Maxwell RSC Instrument, Promega
Maxwell RSC Instrument, Promega
Catalog # PAAS4500
undefined
Maxwell RSC Instrument, Promega
Catalog # PAAS4500
Supplier Number:  AS4500
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Maxwell RSC Instrument
  • Environmentally Preferable:
  • Cat. No.:
    PAAS4500
  • Supplier no.:
    AS4500

Specifications

About this item

The Maxwell Rapid Sample Concentrator (RSC) Instrument is a platform for automated purification of nucleic acid from a range of sample types. Purification methods use sample lysis and binding to paramagnetic particles as the primary separation principle.

  • Automated Nucleic Acid Purification with Integrated Quantitation
  • High-quality nucleic acid purification with minimal steps and less hands-on time
  • Integrated Quantus™ Fluorometer for sample quantitation
  • Intuitive software and touch screen interface

The Maxwell Rapid Sample Concentrator (RSC) Instrument is a platform for automated purification of nucleic acid from a range of sample types. The purification methods use sample lysis and binding to paramagnetic particles as the primary separation principle. Up to 16 samples can be prepared simultaneously in 25-60 minutes, depending on sample type. The Maxwell RSC Instrument is controlled by a graphical user interface running on a tablet PC. The instrument is supplied with a Quantus Fluorometer and integrated software that allows for extracted nucleic acid quantification measurements to be captured in the run report along with sample tracking and method run data. For research use only.

Specifications:
Dimensions: (WxDxH) 330.2 x 345.2 x 299.7 mm (13 x 13.6 x 11.8 in)
Capacity: Up to 16
Molecular Weight: ~85kDa
Processing Time: 30 to 60 min (depending on sample type and method)
UV Lamp: Avg. lifetime approximately 6,000 hrs