Order Entry
United States
ContactUsLinkComponent
Digi-Sense® Pre-Calibrated Humidity and Temperature Indicator, Cole-Parmer
Digi-Sense® Pre-Calibrated Humidity and Temperature Indicator, Cole-Parmer
Catalog # 76183-376
undefined
Digi-Sense® Pre-Calibrated Humidity and Temperature Indicator, Cole-Parmer
Catalog # 76183-376
Supplier Number:  2025030

Specifications

  • Temperature range:
    32...120 °F (0...50 °C)
  • Description:
    Precalibrated Humidity and Temperature Indicator
  • Environmentally Preferable:
  • Cat. No.:
    76183-376
  • Humidity range:
    10 - 90%
  • Temperature accuracy:
    ±1.8 °F (±1.0 °C)

Specifications

About this item

Designed to gauge proper humidity levels.

  • Reliable measurements guaranteed
  • Pre-calibrated to NIST-traceable standards
  • Unique environment comfort indicator
  • Displays both temperature and humidly

Model 20250-30 has built-in temperature and humidity sensors designed for indoor measurements.

Humidity accuracy: ±5% RH; ±8% RH above 80% and below 40% RH

Certifications: NIST-traceable calibration certificate.
Delivery: Includes built-in desktop stand, wall-mounting bracket, one AAA battery, and NIST-traceable calibration report with data supplied by InnoCal.