About this item
Twenty human keratins are resolved with two-dimensional gel electrophoresis into acidic (pI 6.0) subfamilies. This antibody cocktail recognizes acidic (Type I or LMW) and basic (Type II or HMW) cytokeratins, with 67 kDa (CK1); 64 kDa (CK3); 59 kDa (CK4); 58 kDa (CK5); 56 kDa (CK6); 55 kDa (CK7); 52 kDa (CK8); 56.5 kDa (CK10); 53 kDa (CK13); 50 kDa (CK14); 50 kDa (CK15); 48 kDa (CK16); 46 kDa (CK17); 45 kDa (CK18) and 40 kDa (CK19). Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis. KRT-PAN is a broad spectrum anti pan-cytokeratin antibody cocktail, which differentiates epithelial tumors from non-epithelial tumors e.g. squamous vs. adenocarcinoma of the lung, liver carcinoma, breast cancer, and esophageal cancer. It is useful in characterizing the source of various neoplasms and to study the distribution of cytokeratin containing cells in epithelia during normal development and during the development of epithelial neoplasms. This antibody stains cytokeratins present in normal and abnormal human tissues and shows high sensitivity in the recognition of epithelial cells and carcinomas.
CF® dyes are Biotium's next-generation fluorescent dyes. CF®647 is a far-red fluorescent dye (Ex/Em 650/665 nm) with excellent brightness. It also is compatible with super-resolution imaging by STORM.
Type: Primary
Antigen: KRT
Clonality: Monoclonal
Clone: Clone KRT7/760 / KRT17/ 778 / KRT18/834 / KRTL/828 / KRTH/829 / KRT/457
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Chicken, Cow, Dog, Human, Monkey, Mouse, Rabbit, Rat
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 75968-046
- 75968-044
- Antigen Symbol:
- KRT
- KRT
- Antigen Name:
- Cytokeratin
- Cytokeratin
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 647
- CF® 647
- Clone:
- KRT7/760 / KRT17/ 778 / KRT18/834 / KRTL/828 / KRTH/829 / KRT/457
- KRT7/760 / KRT17/ 778 / KRT18/834 / KRTL/828 / KRTH/829 / KRT/457
- Reactivity:
- Rabbit, Monkey, Chicken, Human, Bovine, Dog, Rat, Mouse
- Rabbit, Monkey, Chicken, Human, Bovine, Dog, Rat, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- Unigene ID: 334989 (KRT77) , 654392 (KRT76)
- Unigene ID: 334989 (KRT77) , 654392 (KRT76)
- Isotype:
- IgGs
- IgGs
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0.1 mg/mL
- 0.1 mg/mL
- Storage Buffer:
- PBS/0.1% BSA/0.05% azide
- PBS/0.1% BSA/0.05% azide