iScreen® Urine Drug Control Kit, Alere Toxicology
Supplier: Avantor
Certificates
About this item
iScreen® external controls are use as quality control urine to monitor the precision of laboratory urine toxicology testing procedures for the analytes.
- Liquid solution vials are convenient for any office
- Refrigeration is required
Delivery: Kit includes a positive and negative vial of 5 ml or 20 ml each.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.