GloMax Luminometer Light Plate, Promega
Catalog # PAE6531
Supplier: Promega Corporation
Specifications
- Description:GloMax Luminometer Light Plate
- Environmentally Preferable:
- Cat. No.:PAE6531
- Supplier no.:E6531
Specifications
About this item
The GloMax® 96 Microplate Luminometer Light Plate provides a quick and easy means to verify the performance of the GloMax® 96 Microplate Luminometer. Users can check the sensitivity, reproducibility and linearity.
- Powered by a battery
The Light Plate consists of three highly stable light sources that simulate luminescent samples at signal levels spanning four decades.