To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Magne HaloTag Beads, 20% Slurry, 5ml
- Size:5ml
- Environmentally Preferable:
- Storage Conditions:Store at 2 to 10°C.
- Cat. No.:PAG7282
- Supplier no.:G7282
Specifications
About this item
The Magne HaloTag Beads provide a convenient method to covalently capture HaloTag fusion proteins with magnetic particles for protein pull-downs and purification. The beads offer high binding capacity with low nonspecific protein binding.
- Magnetic Capture of HaloTag(R) Fusion Proteins for Pull-Downs and Purification
- Bind ≥20mg of purified HaloTag(R) fusion protein per ml of particles
- Manual or high-throughput applications
The Magne HaloTag Beads provide a convenient method to covalently capture HaloTag fusion proteins with magnetic particles for protein pull-downs and purification. These magnetic beads offer a high binding capacity (>/=20mg/ml) for purified HaloTag fusion proteins with low nonspecific protein binding. The magnetic handling properties allow streamlined protein purification and eliminate the need for multiple centrifugation steps, facilitating automated applications on robotic platforms. The Magne HaloTag Beads (Cat.# G7281 and G7282) are the recommended replacement for the discontinued HaloLink Magnetic Beads (Cat.# G9311).