Biohazard Bag, Lablemaster
Catalog # 102971-742
Supplier: Label Master
Specifications
- Description:Biohazard 95 kPa pressure bags, XLarge
- Dimensions:13¹/₈×18"
- Environmentally Preferable:
- Cat. No.:102971-742
- Weight:35 lbs.
- Supplier no.:KPA9505
Specifications
About this item
This bag withstand, without leakage, an internal pressure which produces a pressure differential of not less than 95 kPa.
The VonSeal® liquid tight bag has passed the tests as outlined in 173.196(f) of the DOT’s Title 49 CFR and as outlined in Packing Instructions 602 and 650 of the 2001-02 ICAO Technical Instructions for the Safe Transport of Dangerous Goods by Air.
These bags are made with a custom film structure that features a strong adhesive closure and continuous seal, which makes it ideal for critical liquid and substance containment. The permanent adhesive closure withstands a temperature range of –40 °C up to +55 °C, also have a back pocket document pouch.