To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Protein phosphatase 1F
- Clonality:Monoclonal
- Clone:2A9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:9647
- Antigen Synonyms:KIAA0015POPX2CAMKP|Partner of PIX 2|hFEM-2|1F|Ca(2+)/calmodulin-dependent protein kinase phosphatase|protein phosphatase|FEM-2|protein phosphatase 1F|PP2C phosphatase|protein phosphatase 1F (PP2C domain containing)|Protein fem-2 homolog|EC 3.1.3.16|Mg2+/Mn2+ dependent|CaMKPaseCaM-kinase phosphatase
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:PPM1F (NP_055449, 1 a.a. ~ 101 a.a) partial recombinant protein with GST tag.MSSGAPQKSSPMASGAEETPGFLDTLLQDFPALLNPEDPLPWKAPGTVLSQEEVEGELAELAMGFLGSRKAPPPLAAALAHEAVSQLLQTDLSEFRKLPR
- Purification:IgG purified
- Cat. No.:103337-430
- Supplier no.:H00009647-M01
Specifications
About this item
The Protein phosphatase 1F Antibody (2A9) from Novus Biologicals is a mouse monoclonal antibody to Protein phosphatase 1F. This antibody reacts with human. The Protein phosphatase 1F Antibody (2A9) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: Protein phosphatase 1F
Clonality: Monoclonal
Clone: 2A9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human