To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Pleckstrin-2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Environmentally Preferable:
- Gene ID:26499
- Antigen Synonyms:pleckstrin 2|pleckstrin-2|pleckstrin 2 homolog
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:PLEK2 (NP_057529.1, 1 a.a. ~ 353 a.a) full-length human protein.MEDGVLKEGFLVKRGHIVHNWKARWFILRQNTLVYYKLEGGRRVTPPKGRILLDGCTITCPCLEYENRPLLIKLKTQTSTEYFLEACSREERDAWAFEITGAIHAGQPGKVQQLHSLRNSFKLPPHISLHRIVDKMHDSNTGIRSSPNMEQGSTYKKTFLGSSLVDWLISNSFTASRLEAVTLASMLMEENFLRPVGVRSMGAIRSGDLAEQFLDDSTALYTFAESYKKKISPKEEISLSTVELSGTVVKQGYLA
- Purification:Protein A purified
- Cat. No.:103342-352
- Supplier no.:H00026499-B01P
The Pleckstrin-2 Antibody from Novus Biologicals is a mouse polyclonal antibody to Pleckstrin-2. This antibody reacts with human. The Pleckstrin-2 Antibody has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: Pleckstrin-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human