To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:ZNF498
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 μl
- Environmentally Preferable:
- Gene ID:221785
- Antigen Synonyms:Transcriptional repressor RP58|ZBTB1858 kDa repressor protein|C2H2-171|TAZ-1Zinc finger protein C2H2-171|RP58Zinc finger and BTB domain-containing protein 18|TAZ1|Translin-associated zinc finger protein 1|zinc finger protein 238
- Storage Buffer:PBS (pH 7.2) and 40% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against a recombinant protein corresponding to amino acids: PAVNPRDQEMAAGFFTAGSQGLGPFKDMALAFPEEEWRHVTPAQIDCFGEYVEPQDCRVSPGGGSKE
- Purification:Immunogen affinity purified
- Cat. No.:103288-300
- Supplier no.:NBP2-33818
The ZNF498 Antibody from Novus Biologicals is a rabbit polyclonal antibody to ZNF498. This antibody reacts with human. The ZNF498 Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: ZNF498
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human