To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:ZMYND19
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Environmentally Preferable:
- Gene ID:116225
- Antigen Synonyms:zinc finger|MYND domain containing 19|MIZIPmelanin-concentrating hormone receptor 1 interacting zinc-finger protein|zinc finger MYND domain-containing protein 19|RP11-48C7.4|MYND-type containing 19|Melanin-concentrating hormone receptor 1-interacting zinc finger protein|MCH-R1-interacting zinc finger protein
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:ZMYND19 (NP_612471, 1 a.a. ~ 227 a.a) full-length human protein.MTDFKLGIVRLGRVAGKTKYTLIDEQDIPLVESYSFEARMEVDADGNGAKIFAYAFDKNRGRGSGRLLHELLWERHRGGVAPGFQVVHLNAVTVDNRLDNLQLVPWGWRPKAEETSSKQREQSLYWLAIQQLPTDPIEEQFPVLNVTRYYNANGDVVEEEENSCTYYECHYPPCTVIEKQLREFNICGRCQVARYCGSQCQQKDWPAHKKHCRERKRPFQHELEPER
- Purification:Protein A purified
- Cat. No.:103346-366
- Supplier no.:H00116225-B01P
Specifications
About this item
The ZMYND19 Antibody from Novus Biologicals is a mouse polyclonal antibody to ZMYND19. This antibody reacts with human. The ZMYND19 Antibody has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: ZMYND19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human