To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:VPS25
- Clonality:Monoclonal
- Clone:2E5-2B9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:84313
- Antigen Synonyms:MGC10540|ELL-associated protein of 20 kDa|DERP9hVps25|vacuolar protein sorting 25 homolog (S. cerevisiae)|EAP20|EAP20FAP20|vacuolar protein sorting 25 (yeast)|vacuolar protein-sorting-associated protein 25|ESCRT-II complex subunit VPS25|DERP9|Dermal papilla-derived protein 9|FAP20
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:MGC10540 (AAH06282, 1 a.a. ~ 177 a.a) full length recombinant protein with GST tag.MAMSFEWPWQYRFPPFFTLQPNVDTRQKQLAAWCSLVLSFCRLHKQSSMTVMEAQESPLFNNVKLQRKLPVESIQIVLEELRKKGNLEWLDKSKSSFLIMWRRPEEWGKLIYQWVSRSGQNNSVFTLYELTNGEDTEDEEFHGLDEATLLRALQALQQEHKAEIITVSDGRGVKFF
- Purification:IgG purified
- Cat. No.:103345-190
- Supplier no.:H00084313-M01
Specifications
About this item
The VPS25 Antibody (2E5-2B9) from Novus Biologicals is a mouse monoclonal antibody to VPS25. This antibody reacts with human. The VPS25 Antibody (2E5-2B9) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: VPS25
Clonality: Monoclonal
Clone: 2E5-2B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human