To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:USP45
- Clonality:Monoclonal
- Clone:1H2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:85015
- Antigen Synonyms:ubiquitin specific peptidase 45|MGC14793|EC 3.1.2.15|EC 3.4.19.12|Ubiquitin-specific-processing protease 45|Deubiquitinating enzyme 45|ubiquitin carboxyl-terminal hydrolase 45|Ubiquitin thiolesterase 45|ubiquitin thioesterase 45|ubiquitin specific protease 45
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:USP45 (-, 106 a.a. ~ 197 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.FKSSRTEPHCIIINLSTWIIWCYECDEKLSTHCNKKVLAQIVDFLQKHASKTQTSAFSRIMKLCEEKCETDEIQKGGKCRNLSVRGITNLG
- Purification:IgG purified
- Cat. No.:103345-534
- Supplier no.:H00085015-M01
Specifications
About this item
The USP45 Antibody (1H2) from Novus Biologicals is a mouse monoclonal antibody to USP45. This antibody reacts with human, mouse, rat. The USP45 Antibody (1H2) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: USP45
Clonality: Monoclonal
Clone: 1H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse, Rat