To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:UPP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:151531
- Antigen Synonyms:EC 2.4.2.3|UP2|uridine phosphorylase-2|UPASE2|uridine phosphorylase 2|UDRPASE2|UPase 2|UrdPase 2|liver-specific uridine phosphorylase
- Storage Buffer:PBS (pH 7.0) and 20% Glycerol
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human UPP2. The exact sequence is proprietary.
- Purification:Immunogen affinity purified
- Cat. No.:102712-498
- Supplier no.:NBP2-20814
Specifications
About this item
The UPP2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to UPP2. This antibody reacts with human. The UPP2 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: UPP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human