To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:TIM-3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:100 μl
- Environmentally Preferable:
- Gene ID:84868
- Antigen Synonyms:TIM3 T-cell membrane protein 3|TIMD-3|FLJ14428|HAVcr-2|kidney injury molecule-3|T-cell immunoglobulin and mucin domain-containing protein 3|TIMD3KIM-3|Tim-3|hepatitis A virus cellular receptor 2|T cell immunoglobulin mucin 3|T cell immunoglobulin mucin-3|CD366|TIM 3
- Storage Buffer:Whole Rabbit antisera with 50% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:A portion of amino acids 150-200 of human TIM-3 protein were used as the immunogen.
- Purification:Unpurified
- Cat. No.:103358-780
Specifications
About this item
The TIM-3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TIM-3. This antibody reacts with human. The TIM-3 Antibody has been validated for the following applications: ELISA.
Type: Primary
Antigen: TIM-3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human