To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:Th-POK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 ug
- Environmentally Preferable:
- Gene ID:51043
- Antigen Synonyms:THPOK|ZBTB15zinc finger protein 67 homolog (mouse)|T-helper-inducing POZ/Krueppel-like factor|Zinc finger protein 67 homolog|Zinc finger and BTB domain-containing protein 15|zfp-67|ZNF857BDKFZp686G01254|zinc finger and BTB domain-containing protein 7B|zinc finger and BTB domain containing 15|Zinc finger protein Th-POK|zinc finger and BTB domain containing 7B|Zinc finger protein 857B|ZFP67|kruppel-related zinc finger protein hcKrox|hcKroxc-Krox|Krueppel-related zinc finger protein cKrox
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:ZBTB7B (AAH12070.1, 1 a.a. ~ 539 a.a) full-length human protein.
- Purification:Protein G purified
- Cat. No.:103343-606
- Supplier no.:H00051043-B01P
Specifications
About this item
The Th-POK Antibody from Novus Biologicals is a mouse polyclonal antibody to Th-POK. This antibody reacts with human. The Th-POK Antibody has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: Th-POK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human