To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Transglutaminase 1
- Antigen Symbol:TGM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Environmentally Preferable:
- Gene ID:7051
- Antigen Synonyms:Epidermal TGase|transglutaminase 1 (K polypeptide epidermal type I|transglutaminase|LI|keratinocyte|TG(K)|Transglutaminase K|TGASE|ICR2|KTG|transglutaminase-1|TGase-1|LI1EC 2.3.2.13|TGKTGase K|protein-glutamine-gamma-glutamyltransferase)|protein-glutamine gamma-glutamyltransferase K
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:TGM1 (AAH34699, 1 a.a. - 817 a.a.) full-length human protein. MMDGPRSDVGRWGGNPLQPPTTPSPEPEPEPDGRSRRGGGRSFWARCCGCCSCRNAADDDWGPEPSDSRGRGSSSGTRRPGSRGSDSRRPVSRGSGVNAAGDGTIREGMLVVNGVDLLSSRSDQNRREHHTDEYEYDELIVRRGQPFHMLLLLSRTYESSDRITLELLIGNNPEVGKGTHVIIPVGKGGSGGWKAQVVKASGQNLNLRVHTSPNAIIGKFQFTVRTQSDAGEFQLPFDPRNEIYILFNPWCPEDIVYVDHEDWRQEYVLNESGRIYYGTEAQIGERTWNYGQFDHGVLDACLYILDRRGMPYGGRGDPVNVSRVISAMVNSLDDNGVLIGNWSGDYSRGTNPSAWVGSVEILLSYLRTGYSVPYGQCWVFAGVTTTVLRCLGLATRTVTNFNSAHDTDTSLTMDIYFDENMKPLEHLNHDSVWNFHVWNDCWMKRPDLPSGFDGWQVVDATPQETSSGIFCCGPCSVESIKNGLVYMKYDTPFIFAEVNSDKVYWQRQDDGSFKIVYVEEKAIGTLIVTKAISSNMREDITYLYKHPEGSDAERKAVETAAAHGSKPNVYANRGSAEDVAMQVEAQDAVMGQDLMVSVMLINHSSSRRTVKLHLYLSVTFYTGVSGTIFKETKKEVELAPGASDRVTMPVAYKEYRPHLVDQGAMLLNVSGHVKESGQVLAKQHTFRLRTPDLSLTLLGAAVVGQECEVQIVFKNPLPVTLTNVVFRLEGSGLQRPKILNVGDIGGNETVTLRQSFVPVRPGPRQLIASLDSPQLSQVHGVIQVDVAPAPGDGGFFSDAGGDSHLGETIPMASRGGA
- Purification:Protein A purified
- Cat. No.:103333-064
- Supplier no.:H00007051-B02P
Specifications
About this item
The Transglutaminase 1 / TGM1 Antibody from Novus Biologicals is a mouse polyclonal antibody to Transglutaminase 1 / TGM1. This antibody reacts with human. The Transglutaminase 1 / TGM1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: TGM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human