To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:Tbp7
- Clonality:Monoclonal
- Clone:3G8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:5704
- Antigen Synonyms:4,26S proteasome AAA-ATPase subunit RPT3|Proteasome 26S subunit ATPase 4|TBP-7MB67-interacting protein|TBP7MIP224|proteasome (prosome|protease 26S subunit 6|MGC13687,26S protease regulatory subunit 6B|macropain) 26S subunit|MGC23214|ATPase|MGC8570|MB67 interacting protein|S6Tat-binding protein 7
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:PSMC4 (AAH00343, 1 a.a. - 418 a.a.) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MEEIGILVEKAQDEIPALSVSRPQTGLSFLGPEPEDLEDLYSRYKKLQQELEFLEVQEEYIKDEQKNLKKEFLHAQEEVKRIQSIPLVIGQFLEAVDQNTAIVGSTTGSNYYVRILSTIDRELLKPNASVALHKHSNALVDVLPPEADSSIMMLTSDQKPDVMYADIGGMDIQKQEVREAVELPLTHFELYKQIGIDPPRGVLMYGPPGCGKTMLAKAVAHHTTAAFIRVVGSEFVQKYLGEGPRMVRDVFRLAKENAPAIIFIDEIDAIATKRFDAQTGADREVQRILLELLNQMDGFDQNVNVKVIMATNRADTLDPALLRPGRLDRKIEFPLPDRRQKRLIFSTITSKMNLSEEVDLEDYVARPDKISGADINSICQESGMLAVRENRYIVLAKDFEKAYKTVIKKDEQEHEFYK
- Purification:IgG purified
- Cat. No.:103308-504
- Supplier no.:H00005704-M01
The Tbp7 Antibody (3G8) from Novus Biologicals is a mouse monoclonal antibody to Tbp7. This antibody reacts with human. The Tbp7 Antibody (3G8) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: Tbp7
Clonality: Monoclonal
Clone: 3G8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human