To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:TBC1D8
- Clonality:Monoclonal
- Clone:1A12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:11138
- Antigen Synonyms:member 8A|member 8 (with GRAM domain)|AD 3|TBC1 domain family
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:TBC1D8 (NP_008994, 1 a.a. ~ 111 a.a) partial recombinant protein with GST tag.MEQLADVTLRRLLDNEVFDLDPDLQEPSQITKRDLEARAQNEFFRAFFRLPRKEKLHAVVDCSLWTPFSRCHTAGRMFASDSYICFASREDGCCKIILPLREVVSIEKME
- Purification:IgG purified
- Cat. No.:103340-220
- Supplier no.:H00011138-M02
Specifications
About this item
The TBC1D8 Antibody (1A12) from Novus Biologicals is a mouse monoclonal antibody to TBC1D8. This antibody reacts with human. The TBC1D8 Antibody (1A12) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: TBC1D8
Clonality: Monoclonal
Clone: 1A12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human