To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Synphilin-1
- Antigen Symbol:Synphilin-1
- Clonality:Monoclonal
- Clone:2A8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:9627
- Antigen Synonyms:Alpha-synuclein-interacting protein|SYPH1|MGC39814|synphilin-1|Sph1|synuclein|alpha interacting protein
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:SNCAIP (NP_005451, 206 a.a. ~ 310 a.a) partial recombinant protein with GST tag.PFCVLSPVKSPHLRKASAVIHDQHKLSTEETEISPPLVKCGS AYEPENQSKDFLNKTFSDPHGRKVEKTTPDCQLRAFHLQSSA AESKPEEQVSGLNRTSSQGP
- Purification:IgG purified
- Cat. No.:103337-392
- Supplier no.:H00009627-M01
Specifications
About this item
The Synphilin-1 Antibody (2A8) from Novus Biologicals is a mouse monoclonal antibody to Synphilin-1. This antibody reacts with human. The Synphilin-1 Antibody (2A8) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: Synphilin-1
Clonality: Monoclonal
Clone: 2A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human