To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:ST6 Sialyltransferase 2
- Antigen Symbol:ST6GALNAC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:10610
- Antigen Synonyms:SThM|ST6GalNAc II|ST6GalNAcII|STHM(alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminidealpha-2,6-sialyltransferase|sialyltransferase 7((alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminidealpha-2,6-sialyltransferase)|SAITL1|alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase 2|sialyltransferase-like 1|Sialyltransferase 7B|ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminidealpha-2,6-sialyltransferase 2|GalNAc alpha-2,6-sialyltransferase II|sialyltransferase 7((alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminidealpha-2,6-sialyltransferase) B|sialyltransferase 7((alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide B|EC 2.4.99.-|FLJ45660|SIAT7-B|(alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide B|SIAT7B|SIAT7|SIATL1|ST6GalNAII
- Storage Buffer:0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human ST6GALNAC2. The exact sequence is proprietary.
- Purification:Immunogen affinity purified
- Cat. No.:102711-858
- Supplier no.:NBP2-20494
Specifications
About this item
The ST6 Sialyltransferase 2 / ST6GALNAC2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to ST6 Sialyltransferase 2 / ST6GALNAC2. This antibody reacts with human. The ST6 Sialyltransferase 2 / ST6GALNAC2 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: ST6GALNAC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human