To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:SRGN
- Clonality:Monoclonal
- Clone:1D8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:5552
- Antigen Synonyms:secretory granule proteoglycan 1|secretory granule|P.PG|proteoglycan protein core for mast cell secretory granule|MGC9289|PRGhematopoetic proteoglycan core peptide|serglycin|Secretory granule proteoglycan core protein|FLJ12930|secretory granule proteoglycan core peptide|serglycin proteoglycan|Platelet proteoglycan core protein|Hematopoetic proteoglycan core protein|platelet proteoglycan protein core|PPG|proteoglycan 1|PRG1p.PG
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:SRGN (AAH15516, 1 a.a. - 158 a.a.) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MMQKLLKCSRLVLALALILVLESSVQGYPTQRARYQWVRCNPDSNSANCLEEKGPMFELLPGESNKIPRLRTDLFPKTRIQDLNRIFPLSEDYSGSGFGSGSGSGSGSGSGFLTEMEQDYQLVDESDAFHDNLRSLDRNLPSDSQDLGQHGLEEDFML
- Purification:IgG purified
- Cat. No.:103308-024
- Supplier no.:H00005552-M03
Specifications
About this item
The SRGN Antibody (1D8) from Novus Biologicals is a mouse monoclonal antibody to SRGN. This antibody reacts with human. The SRGN Antibody (1D8) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: SRGN
Clonality: Monoclonal
Clone: 1D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human