To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:RUVBL2
- Clonality:Monoclonal
- Clone:3C6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:10856
- Antigen Synonyms:51 kDa erythrocyte cytosolic protein|REPTIN|Reptin52|RuvB (E coli homolog)-like 2|TIP60-associated protein 54-beta|RVB2|TBP-interacting protein|INO80 complex subunit J|51-KD|EC 3.6.4.12,48 kDa TBP-interacting protein|TIP48ECP-51|erythrocyte cytosolic protein|48 kDa TATA box-binding protein-interacting protein|ruvB-like 2|TIH2|TIP49b|Reptin 52|RuvB-like 2 (E. coli)|EC 3.6.1|INO80JTAP54-beta|ECP51TIP49B|Repressing pontin 52|48-KD
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:RUVBL2 (AAH00428, 1 a.a. ~ 464 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.MATVTATTKVPEIRDVTRIERIGAHSHIRGLGLDDALEPRQASQGMVGQLAARRAAGVVLEMIREGKIAGRAVLIAGQPGTGKTAIAMGMAQAPGPDTPFTAIAGSEIFSLEMSKTEALTQAFRRSIGVRIKEETEIIEGEVVEIQIDRPATGTGSKVGKLTLKTTEMETIYDLGTKMIESLTKDKVQAGDVITIDKATGKISKLGRSFTRARDYDAMGSQTKFVQCPDGELQKRKEVVHTVSLHEIDVINSRTQ
- Purification:Protein A purified
- Cat. No.:103339-672
- Supplier no.:H00010856-M01
Specifications
About this item
The RUVBL2 Antibody (3C6) from Novus Biologicals is a mouse monoclonal antibody to RUVBL2. This antibody reacts with human. The RUVBL2 Antibody (3C6) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: RUVBL2
Clonality: Monoclonal
Clone: 3C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human