To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:RTVP-1/GLIPR1
- Clonality:Monoclonal
- Clone:8D9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:11010
- Antigen Synonyms:and pathogenesis proteins 1|GliPR|GLIPRglioma pathogenesis-related protein 1|vespid|GLI pathogenesis-related 1|related to testis-specific|RTVP1CRISP7|Protein RTVP-1|testes-specific vespid and pathogenesis protein 1|GLI pathogenesis-related 1 (glioma)|GliPR 1
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:GLIPR1 (NP_006842, 23 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Purification:IgG purified
- Cat. No.:103339-948
- Supplier no.:H00011010-M04
Specifications
About this item
The RTVP-1 / GLIPR1 Antibody (8D9) from Novus Biologicals is a mouse monoclonal antibody to RTVP-1 / GLIPR1. This antibody reacts with human. The RTVP-1 / GLIPR1 Antibody (8D9) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: RTVP-1/GLIPR1
Clonality: Monoclonal
Clone: 8D9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human