To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:RGM-C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:148738
- Antigen Synonyms:repulsive guidance molecule c|hemochromatosis type 2 (juvenile)|hemojuvelin|HFE2AMGC23953|RGMCRGM domain family member C|HJVHemochromatosis type 2 protein|haemojuvelin|JH
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:HFE2 (NP_660320.3, 1 a.a. ~ 313 a.a) full-length human protein.MIQHNCSRQGPTAPPPPRGPALPGAGSGLPAPDPCDYEGRFSRLHGRPPGFLHCASFGDPHVRSFHHHFHTCRVQGAWPLLDNDFLFVQATSSPMALGANATATRKLTIIFKNMQECIDQKVYQAEVDNLPVAFEDGSINGGDRPGGSSLSIQTANPGNHVEIQAAYIGTTIIIRQTAGQLSFSIKVAEDVAMAFSAEQDLQLCVGGCPPSQRLSRSERNRRGAITIDTARRLCKEGLPVEDAYFHSCVFDVLIS
- Purification:Protein A purified
- Cat. No.:103347-212
- Supplier no.:H00148738-D01P
Specifications
About this item
The RGM-C / Hemojuvelin Antibody from Novus Biologicals is a rabbit polyclonal antibody to RGM-C / Hemojuvelin. This antibody reacts with human, mouse. The RGM-C / Hemojuvelin Antibody has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: RGM-C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse