To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:PPP2R2C
- Clonality:Monoclonal
- Clone:6D1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:5522
- Antigen Synonyms:gamma|subunit B|regulatory subunit B|R2-gamma isoform|gamma isoform|regulatory subunit B (PR 52)|gammaisoform|phosphoprotein phosphatase 2A BR gamma regulatory chain|PP2A subunit B isoform PR55-gamma|IMYPNO1|protein phosphatase 2A1 B gamma subunit|PP2A|B55-GAMMA|B55-gamma isoform|55 KDA regulatory subunit B|PP2A subunit B isoform gamma|B-gamma isoform|PR55-gamma isoform|PP2A subunit B isoform B55-gamma|PR55G|protein phosphatase 2 (formerly 2A)|PP2A subunit B isoform R2-gamma|serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B gammaisoform|serine/threonine protein phosphatase 2A|protein phosphatase 2
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:PPP2R2C (NP_065149, 1 a.a. - 110 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MGEDTDTRKINHSFLRDHSYVTEADIISTVEFNHTGELLATGDKGGRVVIFQREPESKNAPHSQGEYDVYSTFQSHEPEFDYLKSLEIEEKINKIKWLPQQNAAHSLLST
- Purification:IgG purified
- Cat. No.:103307-964
- Supplier no.:H00005522-M01
Specifications
About this item
The PPP2R2C Antibody (6D1) from Novus Biologicals is a mouse monoclonal antibody to PPP2R2C. This antibody reacts with human. The PPP2R2C Antibody (6D1) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: PPP2R2C
Clonality: Monoclonal
Clone: 6D1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human